Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Neuroligin 4Y/NLGN4Y Protein

Catalog No. 9119NL050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-19N-L050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 91-19N-L050 Supplier R&D Systems™ Supplier No. 9119NL050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Neuroligin 4Y/NLGN4Y Protein is derived from CHO. The Recombinant Human Neuroligin 4Y/NLGN4Y Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q8NFZ3
For Use With (Application) Bioactivity
Gene ID (Entrez) 22829
Molecular Weight (g/mol) Observed M.W.: 90-105kDa, reducing conditions, Theoretical M.W.: 72kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived human Neuroligin 4Y/NLGN4Y protein Gln42-Val677,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias neuroligin 4, Y-linked, Neuroligin Y, neuroligin-4, Y-linked, NLGN4Y, Y linked
Common Name Neuroligin 4Y/NLGN4Y
Biological Activity Measured by its ability to enhance neurite outgrowth of dissociated E13 chick embryonic dorsal root ganglia (DRG) neurons. Able to significantly enhance neurite outgrowth when immobilized at 3-12μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Gln42-Val677, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.