Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human NF-L Protein
SDP

Catalog No. 9715NF050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
97-15N-F050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 97-15N-F050 Supplier R&D Systems™ Supplier No. 9715NF050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Derived from E. coli.

Specifications

Accession Number P07196
For Use With (Application) Flow Cytometry
Gene ID (Entrez) 4747
Molecular Weight (g/mol) 41 kDa, reducing conditions
Quantity 50 μg
Source E. coli-derived human NF-L protein Ser2-Asp338, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration < 0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500 μg/mL in 4 mM HCl
Buffer Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA
Purity or Quality Grade > 95%, by SDS-PAGE under reducing conditions and visualized by silver Stain
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.