Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human NFATC2 Protein

Catalog No. 9716NA050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
97-16N-A050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 97-16N-A050 Supplier R&D Systems™ Supplier No. 9716NA050
Add to Cart
Add to Cart
This item is not returnable. View return policy

R&D Systems™ Recombinant Human NFATC2 Protein CF is a member of the calcium-regulated family of transcription factors that regulate gene expression in lymphocytes and control cardiac valve formation.

The Recombinant Human NFATC2 Protein is derived from E. coli.

Specifications

Accession Number Q13469-1
For Use With (Application) Flow Cytometry
Formulation Lyophilized from a 0.2μm filtered solution in HCl.
Gene ID (Entrez) 4773
Molecular Weight (g/mol) 13 kDa
Name NFAT pre-existing subunit, NFAT transcription complex, preexisting component, NFAT1, NFAT1nuclear factor of activated T-cells, cytoplasmic 2, NFATc2, NF-ATc2, NFATp, NF-ATp, NFATPnuclear factor of activated T-cells, preexisting component, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2, preexisting nuclear factor of activated T-cells 2, T cell transcription factor NFAT1, T-cell transcription factor NFAT1
Quantity 50 μg
Source E. coli-derived human NFATC2 protein His575-Pro679,with an N-terminal Met and a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500μg/mL in 4 mM HCl..
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.