Learn More
R&D Systems™ Recombinant Human NKp30 Fc Chimera Avi-tag Protein
Description
Specifications
Specifications
| Accession Number | Q14931.1 |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 259197 |
| Molecular Weight (g/mol) | M.W.-observed: 53-63 kDa, under reducing conditions., M.W.-theroretical: 40 kDa (monomer) |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human NKp30/NCR3 protein Human NKp30 (Leu19-Thr138) (C-terminus) Accession # Q14931.1 IEGRMD Human IgG1 (Pro100-Lys330) Avi-tag (N-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | Activating natural killer receptor p30, CD337, CD337 antigen, CD337activating NK-A1 receptor, LY117,1C7NK-p30, lymphocyte antigen 117, natural cytotoxicity triggering receptor 3, Natural killer cell p30-related protein, NCR3, NKP30, NKp30MALS |
| Conjugate | Biotin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.