Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Noggin Fc Chimera Protein

Catalog No. 3344NG050
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3344NG050 50 μg
33-44N-G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 3344NG050 Supplier R&D Systems™ Supplier No. 3344NG050
Add to Cart
Add to Cart
This item is not returnable. View return policy

A New and Improved Noggin is Now Available! The new protein has ∼2 fold better activity, higher purity, and no tag! Earn double rewards for reviews

Measured by its ability to inhibit BMP-4-induced alkaline phosphatase production by ATDC5 mouse chondrogenic cells. The ED50 for this effect is 0.100-0.400 µg/mL in the presence of 30 ng/mL of Recombinant Human BMP-4 (Catalog # 314-BP ).

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 9241
Molecular Weight (g/mol) 49.6 kDa (monomer)
Name Noggin Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human Noggin protein, (N-terminus) Human Noggin (Gln28-Cys232) Accession &Num; Q13253 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Gene Alias NOG, noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1, SYNS1A
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.