Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Noggin Fc Chimera Protein

Catalog No. 3344NG100
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-44N-G100 100 μg
3344NG050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-44N-G100 Supplier R&D Systems™ Supplier No. 3344NG100
Add to Cart
Add to Cart
This item is not returnable. View return policy

A New and Improved Noggin is Now Available! The new protein has ∼2 fold better activity, higher purity, and no tag! Earn double rewards for reviews

Measured by its ability to inhibit BMP-4-induced alkaline phosphatase production by ATDC5 mouse chondrogenic cells. The ED50 for this effect is 0.100-0.400 µg/mL in the presence of 30 ng/mL of Recombinant Human BMP-4 (Catalog # 314-BP ).

Specifications

Accession Number Q13253
For Use With (Application) Bioactivity
Gene ID (Entrez) 9241
Molecular Weight (g/mol) 49.6 kDa (monomer)
Name NOG, noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1, SYNS1A
Quantity 100 μg
Source

Mouse myeloma cell line, NS0-derived human Noggin protein

Human Noggin (Gln28-Cys232) Accession # Q13253IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Gene Symbol NOG
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.