Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human NPDC-1 Protein

Catalog No. 9724NP050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9724NP050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 9724NP050 Supplier R&D Systems™ Supplier No. 9724NP050
Add to Cart
Add to Cart
This item is not returnable. View return policy

R&D Systems™ Recombinant Human NPDC-1 Protein CF is a protein that is mainly expressed in lung and neural tissues and whose expression is developmentally regulated and persists in the adult.

The Recombinant Human NPDC-1 Protein is derived from NS0.

Specifications

Accession Number Q9NQX5
For Use With (Application) Flow Cytometry
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 56654
Molecular Weight (g/mol) 16 kDa
Name CAB-, CAB1, CAB-1, CAB1CAB, DKFZp586J0523, neural proliferation differentiation and control protein 1, neural proliferation, differentiation and control, 1, NPDC1, NPDC-1
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human NPDC-1 protein Gly35-Asp181,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 250μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.