Learn More
R&D Systems™ Recombinant Human NRG1-beta 1/HRG1-beta 1 ECD Protein
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
| Gene ID (Entrez) | 3084 |
| Molecular Weight (g/mol) | 26.9 kDa |
| Name | Neuregulin-1 beta 1/NRG1 beta 1 Extracellular Domain |
| Quantity | 50 μg |
| Source | E. coli-derived human Neuregulin-1 beta 1/NRG1 beta 1 protein Ser2-Lys246 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Product Line | R&D Systems |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.