Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human NRG1-beta 1/HRG1-beta 1 Fc Protein, CF

Catalog No. 11610NR050
Change view
Click to view available options
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-610-NR050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-610-NR050 Supplier R&D Systems™ Supplier No. 11610NR050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Fc Chimera

Measured in a serum-free cell proliferation assay using MCF-7 human breast cancer cells. Karey, K.P. et al. (1988) Cancer Research 48:4083. The ED50 for this effect is 2.00-20.0 ng/mL.

Specifications

For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3084
Molecular Weight (g/mol) M.W.-observed: 66-73 kDa, under reducing conditions, M.W.-theroretical: 52 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Neuregulin-1 beta 1/NRG1 beta 1 protein Human NRG1-beta1 (Ser 20-Lys 246) (N-terminus) Accession # NP_039250.2 GGIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, MST131, MSTP131, NDF, neuregulin 1, Neuregulin1 beta 1, NRG1-IT2, SMDF
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 500μg/mL in sterile water.
Form Lyophilized
Protein Neuregulin-1 beta 1/NRG1 beta 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.