Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human NUBP2 His Protein
SDP

Catalog No. NBP25190405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25190405 0.05 mg
NBP25190425 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25190405 Supplier Novus Biologicals™ Supplier No. NBP2519040.05MG
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-271 of Human NUBP2 The Recombinant Human NUBP2 Protein is derived from E. coli. The Recombinant Human NUBP2 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation Phosphate buffered saline (pH7.4) containing 50% glycerol, 1mM DTT
Gene ID (Entrez) 10101
Molecular Weight (g/mol) TMW: 31.2kDa
Quantity 0.05 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias CFD1C447E6.1 (nucleotide binding protein 1 (E.coli MinD like) ), cytosolic Fe-S cluster assembly factor NUBP2, homolog of yeast cytosolic Fe-S cluster deficient 1, NBP 2, NUBP1, nucleotide binding protein 2 (E.coli MinD like), nucleotide binding protein 2 (MinD homolog, E. coli), Nucleotide-binding protein 2
Common Name NUBP2
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.