Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human O-GlcNAcTransferase/OGT Protein

Catalog No. 8446GT050
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
84-46G-T050 50 μg
84-46G-T010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 84-46G-T050 Supplier R&D Systems™ Supplier No. 8446GT050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to transfer GlcNAc from UDP-GclNAc to peptide OGT substrate from AnaSpec, Inc. The specific activity is >25 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number O15294
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and TCEP.
Gene ID (Entrez) 8473
Molecular Weight (g/mol) M.W. (Observed): 70 kDa, reducing conditions; M.W. (theoretical): 81 kDa
Name O-GlcNAc Transferase/OGT
Quality Control Testing SDS-PAGE: 70kDa, reducing conditions
Quantity 50 μg
Source E. coli-derived human O-GlcNAc Transferase/OGT protein Cys323-Glu1041, with C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Regulatory Status RUO
Termination Cys323
Without Additives Carrier Protein
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias EC 2.4.1, EC 2.4.1.255 FLJ23071, HRNT1, MGC22921, O-GLCNAC, OGlcNAc Transferase, O-GlcNAc transferase p110 subunit, O-GlcNAc transferase subunit p110, OGT, O-linked N-acetylglucosamine (GlcNAc) transferase(UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase), O-linked N-acetylglucosamine transferase 110 kDa subunit, UDP-N-acetylglucosamine--peptide N-acetylglucosaminyltransferase 110 kDasubunit, uridinediphospho-N-acetylglucosamine:polypeptide beta-N-acetylglucosaminyltransferase
Gene Symbol OGT
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >80%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein O-GlcNAc Transferase p110 subunit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.