Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Octa-Ub/Ub8 WT Chains (K63-linked), CF

Catalog No. UC318B025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
UC318B025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. UC318B025 Supplier R&D Systems™ Supplier No. UC318B025
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Enzyme Activity

The Recombinant Human Octa-Ub/Ub8 WT Chains (K63-linked) is derived from E. coli. The Recombinant Human Octa-Ub/Ub8 WT Chains (K63-linked) has been validated for the following applications: Bioactivity, Enzyme Activity.

Specifications

Accession Number P0CG47
For Use With (Application) Bioactivity, Enzyme Activity
Gene ID (Entrez) 7314
Molecular Weight (g/mol) 68 kDa
Quantity 25 μg
Source E. coli-derived human Octa-Ubiquitin protein
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening.
Regulatory Status RUO
Specific Reactivity Octa-Ubiquitin
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Buffer 1 mg/mL (15 μm) in sterile, deionized water
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.