Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Oncostatin M (HEK-293-expressed)

Catalog No. 8475OM010CF
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
84-75O-M010CF 10 μg
84-75O-M050CF 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 84-75O-M010CF Supplier R&D Systems™ Supplier No. 8475OM010/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Oncostatin M (HEK-293-expressed) is derived from HEK293. The Recombinant Human Oncostatin M (HEK-293-expressed) has been validated for the following applications: Bioactivity.

Specifications

Accession Number P13725
For Use With (Application) Bioactivity
Gene ID (Entrez) 5008
Molecular Weight (g/mol) Observed M.W.: 27-35kDa, reducing conditions, Theoretical M.W.: 22kDa
Quantity 10 μg
Source Human embryonic kidney cell,HEK293-derived human Oncostatin M/OSM protein Ala26-Arg221
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias MGC20461, oncostatin M, oncostatin-M
Common Name Oncostatin M/OSM
Gene Symbol OSM
Biological Activity Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is typically 0.05-0.3ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Ala26-Arg221
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.