Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human OR1A1 GST (N-Term) Protein
SDP

Catalog No. NB170397 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170397 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB170397 Supplier Novus Biologicals™ Supplier No. H00008383P012UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.

Specifications

For Use With (Application) ELISA, Immunoaffinity Purification, Protein Array, Western Blot
Formulation 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 8383
Molecular Weight (g/mol) M.W. Theoretical: 61 kDa
Quantity 2 μg
Storage Requirements Store at -80C. Avoid freeze-thaw cycles.
Gene Alias olfactory receptor 1A1, Olfactory receptor OR17-11, olfactory receptor, family 1, subfamily A, member 1, OR17-7Olfactory receptor 17-7
Gene Symbol OR1A1
Research Category GPCR, Neuroscience, Signal Transduction
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein OR1A1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.