Learn More
Novus Biologicals™ Recombinant Human Ornithine Carbamoyltransferase His Protein
Shop All R&D Systems Products

Description
Specifications
Specifications
| Accession Number | P00480 |
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. In 20mM MES buffer (pH 6.0) containing 2mM DTT, 10% glycerol, 100mM NaCl |
| Gene ID (Entrez) | 5009 |
| Molecular Weight (g/mol) | 38.9kDa |
| Name | Ornithine Carbamoyltransferase Protein |
| Purification Method | SDS-PAGE |
| Storage Requirements | 4°C short term, −20°C long term. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Purity or Quality Grade | >90% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.