Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Osteoprotegerin/TNFRSF11B Protein

Catalog No. 185OS025CF
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
185OS025CF Lyophilized from a 0.2 μm filtered solution in PBS.
185OS025 Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 185OS025CF Supplier R&D Systems™ Supplier No. 185OS025/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

A New and Improved rh OPG is Now Available! The new protein has ∼2 fold better activity!

Measured by its ability to inhibit TRAIL-mediated cytotoxicity using L-929 mouse fibroblast cells treated with TRAIL. The ED50 for this effect is 8-24 ng/mL.

Specifications

Accession Number AAB53709.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 4982
Molecular Weight (g/mol) M.W. (Observed): 55 kDa, reducing conditions; M.W. (theoretical): 43.5 kDa (monomer)
Name Osteoprotegerin/TNFRSF11B
Quantity 25 μg
Source Mouse myeloma cell line, NS0-derived human Osteoprotegerin/TNFRSF11B protein Glu22-Leu401
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol TNFRSF11B
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Osteoprotegerin/TNFRSF11B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.