Learn More
R&D Systems™ Recombinant Human OX40 Ligand/TNFSF4 Protein
Description
Specifications
Specifications
| Accession Number | Q6FGS4 |
| For Use With (Application) | Bioactivity, Binding Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 7292 |
| Molecular Weight (g/mol) | M.W.-observed: 28-34 kDa, reducing conditions, M.W.-theroretical: 18 kDa |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line, NS0-derived human OX40 Ligand/TNFSF4 protein MHHHHHHHHHH GGGSGGGSGGGS IEGR Human OX40 Ligand (Gln51-Leu183) (N-terminus) Accession # Q6FGS4 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, O x 40 antigen ligand, O x 40 ligand, O x 40 L, OX-40 L, O x 4 OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34 kD), tax-transcriptionally activated glycoprotein 1, 34 kD, TNFSF4, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.