Learn More
R&D Systems™ Recombinant Human OX40/TNFRSF4 Fc Alexa Fluor™ 488 Protein
Description
Specifications
Specifications
| Accession Number | P43489.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein |
| Gene ID (Entrez) | 7293 |
| Molecular Weight (g/mol) | M.W.-observed: 70 kDa, reducing conditions, M.W.-theroretical: 46.7 kDa (monomer) Labeled with Alexa Fluor™ 488 via amines Excitation Wavelength: 488 nm Emission Wavelength: 515-545 nm |
| Quantity | 20 μg |
| Source | Mouse myeloma cell line, NS0-derived human O x 40/TNFRSF4 protein Human O x 40 (Leu29-Ala216) (N-terminus) Accession # P43489.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | ACT-135, ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, Ly-70, O x 40 cell surface antigen, O x 40 homologue, O x 40 L receptor, O x 40 lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, TNFRSF4, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, Txgp1, TXGP1L, TXGP1LO x 40 antigen |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.