Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Pappalysin-1/PAPP-A (aa 82-1627)

Catalog No. 2487ZNF020
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2487ZNF020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 2487ZNF020 Supplier R&D Systems™ Supplier No. 2487ZNF020
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to cleave IGFBP-5. Recombinant Human Pappalysin-1/PAPP-A will cleave >50% of Recombinant Human IGFBP-5 (Catalog # 875-B5 ), as measured under the described conditions.

Specifications

Accession Number Q13219
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl, CaCl2, and CHAPS.
Gene ID (Entrez) 5069
Molecular Weight (g/mol) M.W. (Observed): 139-238 kDa, three bands, reducing conditions; M.W. (theoretical): 173 kDa
Name Pappalysin-1/PAPP-A aa 82-1627
Quantity 20 μg
Source Mouse myeloma cell line, NS0-derived human Pappalysin-1/PAPP-A protein Glu82-Gly1627 (S1224Y), with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol PAPPA
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein Pappalysin-1/PAPP-A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.