Learn More
R&D Systems™ Recombinant Human PD-L1/B7-H1 His Alexa Fluor™ 488 Protein
Description
Specifications
Specifications
| Accession Number | Q9NZQ7.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS and NaCl with BSA as a carrier protein. |
| Gene ID (Entrez) | 29126 |
| Molecular Weight (g/mol) | M.W.-observed: 31-41 kDa, under reducing conditions., M.W.-theroretical: 26 kDa |
| Quantity | 20 μg |
| Source | Human embryonic kidney cell, HEK293-derived human PD-L1/B7-H1 protein Phe19-Thr239, with a C-terminal 6-His tag |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.