Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human PD-L1/B7-H1 His-tag Protein

Catalog No. 9049B7100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-49B-7100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-49B-7100 Supplier R&D Systems™ Supplier No. 9049B7100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Measured by its binding ability in a functional ELISA. When Recombinant Human PD-L1/B7-H1 is immobilized at 0.025 μg/mL (100 μL/well), the concentration of Recombinant Human PD-1 Fc Chimera (Catalog # 1086-PD ) that produces 50% of the optimal binding response is approximately 0.05-0.3 μg/mL.

Specifications

Accession Number Q9NZQ7
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 29126
Molecular Weight (g/mol) M.W. (Observed): 31-41 kDa, reducing conditions; M.W. (theoretical): 26 kDa
Quantity 100 μg
Source Human embryonic kidney cell, HEK293-derived human PD-L1/B7-H1 protein Phe19-Thr239, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1
Common Name B7-H1/PD-L1/CD274
Gene Symbol CD274
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human B7-H1 is immobilized at 0.025μg/mL (100μL/well), the concentration of Recombinant Human PD-1 Fc Chimera that produces 50% of the optimal binding response is approximately 0.05-0.3μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in PBS.
Expression System Human embryonic kidney cell, HEK293-derived Phe19-Thr239, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PD-L1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.