Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human PD-L2/B7-DC Fc Alexa Fluor™ 647 Protein

Catalog No. AFR1224020
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AFR1224020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AFR1224020 Supplier R&D Systems™ Supplier No. AFR1224020
Add to Cart
Add to Cart
This item is not returnable. View return policy
Measured by flow cytometry for its ability to bind anti-human PD-L2/B7-DC Monoclonal Antibody conjugated beads. The concentration of Recombinant Human PD-L2/B7-DC Chimera Alexa Fluor™ 647 (Catalog # AFR1224) that produces 50% of the binding response is 8.00-80.0 ng/mL.

Specifications

Accession Number Q9BQ51.2
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 80380
Molecular Weight (g/mol) M.W.-observed: 70-80 kDa, under reducing conditions., M.W.-theroretical: 49 kDa (monomer)
Quantity 20 μg
Source Mouse myeloma cell line, NS0-derived human PD-L2/B7-DC protein Human PD-L2 (Leu20-Pro219) (C-terminus) Accession # Q9BQ51.2 IEGRMD Human IgG1 (Pro100-Lys330) (N-terminus)
Storage Requirements Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, Butyrophilin-like Protein, CD273, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDCD1LG2, PDL2, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2
Conjugate Alexa Fluor 647
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PD-L2/B7-DC/PDCD1LG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.