Learn More
R&D Systems™ Recombinant Human PD-L2/B7-DC His Alexa Fluor™ 488 Protein
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Accession Number | Q9BQ51.2 |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 80380 |
| Molecular Weight (g/mol) | M.W.-observed: 29-47 kDa, under reducing conditions., M.W.-theroretical: 23 kDa |
| Quantity | 20 μg |
| Source | Human embryonic kidney cell, HEK293-derived human PD-L2/B7-DC protein Leu20-Pro219, with a C-terminal 6-His tag |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, Butyrophilin-like Protein, CD273, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDCD1LG2, PDL2, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2 |
| Conjugate | Alexa Fluor 488 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.