Learn More
R&D Systems™ Recombinant Human PDGF-AA Protein
Description
Specifications
Specifications
| Accession Number | P04085 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA. *1 mg pack size (01M) is supplied as a 0.2 μm filtered solution in Acetonitrile and TFA. |
| Gene ID (Entrez) | 5154 |
| Molecular Weight (g/mol) | M.W. theroretical: 14 kDa (monomer) |
| Quantity | 25 μg |
| Source | <EM>E. coli</EM>-derived human PDGF-AA protein Ser87-Thr211, with and without an N-terminal Met |
| Storage Requirements | Manual defrost freezer; Avoid repeated freeze-thaw cycles; 12 mo. from receipt, -20 to -70°C as supplied; 1 mo, 2 to 8°C under sterile conditions after reconstitution; 3 mo, -20 to -70°C under sterile conditions after reconstitution |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.01 EU per 1 μg of the protein by the LAL method. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.