Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human PDGF-AB (single chain) Protein, CF

Catalog No. 11471AB010
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-471-AB010 10 μg
11-471-AB01M 1 mg
11-471-AB050 50 μg
11-471-AB100 100 μg
11-471-AB250 250 μg
11-471-AB500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 11-471-AB010 Supplier R&D Systems™ Supplier No. 11471AB010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is 2.00-20.0 ng/mL.

Specifications

Accession Number NP_001382295.1 & P01127.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose.
Gene ID (Entrez) 5154
Molecular Weight (g/mol) MolecularWeight-observed: 28-37 kDa, under reducing conditions., MolecularWeight-theroretical: 26 kDa
Quantity 10 μg
Source Human embryonic kidney cell, HEK293-derived human PDGF-AB protein Human PDGFA (Ser87-Arg196) (Cys123Ala, Cys132Ala) (N-terminus) Accession # NP_001382295.1 linker Human PDGFB (Ser82-Thr190) (Cys123Ala, Cys132Ala) (N-terminus) Accession # P01127.1 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Reconstitution Reconstitute 10 μg size at 100 μg/mL in 4 mM HCl. Reconstitute all the other sizes at 250 μg/mL in 4 mM HCl.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PDGF-AB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.