Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human PDGF-BB, Biotinylated Protein

Catalog No. BT220010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BT220010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BT220010 Supplier R&D Systems™ Supplier No. BT220010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human PDGF-BB, Biotinylated Protein is derived from E. coli. The Recombinant Human PDGF-BB, Biotinylated Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q6FHE7
For Use With (Application) Bioactivity
Gene ID (Entrez) 5155
Molecular Weight (g/mol) Observed Molecular Wt: 12kDa, reducing conditions, Predicted Molecular Wt.: 12kDa (unlabeled)
Quantity 10 μg
Source E. coli-derived human PDGF-BB protein Ser82-Thr190
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer, biotinylated protein via amines
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Common Name PDGF-BB
Gene Symbol PDGFB
Biological Activity Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is typically 1.5-6ng/mL.
Conjugate Biotin
Species Human
Recombinant Recombinant
Expression System E. coli-derived Ser82-Thr190
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.