Learn More
R&D Systems™ Recombinant Human PDGF R alpha His-tag Avi-tag Protein
Description
Specifications
Specifications
| Accession Number | P16234.1 |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 5156 |
| Molecular Weight (g/mol) | Observed: 82-108 kDa, under reducing conditions; Theroretical: 59 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human PDGF R alpha protein Human PDGF R alpha (Gln24-Glu524) Accession # P16234.1(N-terminus) HHHHHH Avi-tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Biotin |
| Reconstitution | Reconstitute at 500 μg/mL in PBS. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.