Learn More
Proteintech Group Inc Recombinant Human PDGFbb 10ug
SDP
Supplier:  Proteintech Group Inc HZ130810UG
Encompass_Preferred
Animal-free Recombinant Human PDGFbb is a growth factor that promotes blood vessel formation, mitogenesis, chemotaxis, etc. The PDGF family members (four homodimers and one heterodimer) are secreted, disulfide-linked dimeric glycoproteins that regulate their cellular functions through interaction with PDGFR receptors. PDGFbb is synthesized, stored, and released by alpha granules of platelets. It is also known as PDGF-2, becaplermin, or GDGF. Dysregulation in PDGF signaling has been shown to be associated with tumorigenesis and progression of cancer. Recombinant PDGFbb is used in treatment of chronic ulcers and to speed healing in surgical procedures.
Catalog No. 50-197-3974
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.