Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human PDHX His Protein
SDP

Catalog No. p-7104843 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25944002 0.02 mg
NBP25194401 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25944002 Supplier Novus Biologicals™ Supplier No. NBP2519440.02MG
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 54-501 of Human PDHX The Recombinant Human PDHX Protein is derived from E. coli. The Recombinant Human PDHX Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.25mg/mL
For Use With (Application) SDS-PAGE
Formulation Phosphate buffer saline (pH 7.4) containing 20% glycerol, 1mM DTT
Gene ID (Entrez) 8050
Molecular Weight (g/mol) TMW: 50.4kDa
Quantity 0.02 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias Dihydrolipoamide dehydrogenase-binding protein of pyruvate dehydrogenasecomplex, DLDBP, E3-binding protein, E3BPpyruvate dehydrogenase complex, E3-binding protein subunit, OPDX, PDX1Lipoyl-containing pyruvate dehydrogenase complex component X, proXpyruvate dehydrogenase complex, lipoyl-containing component X, pyruvate dehydrogenase complex, component X, pyruvate dehydrogenase protein X component, mitochondrial
Common Name PDHX
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.