Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Pepsinogen A5/PGA5 Protein

Catalog No. 8457AS020
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
84-57A-S020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 84-57A-S020 Supplier R&D Systems™ Supplier No. 8457AS020
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to cleave the fluorogenic peptide substrate, Mca-RPKPVE-Nval-WRK(Dnp)-NH2 (Catalog # ES002 ). The specific activity is >15,500 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number P0DJD9
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 5222
Molecular Weight (g/mol) M.W. (Observed): 39-51 kDa, reducing conditions; M.W. (theoretical): 41 kDa
Quantity 20 μg
Source Human embryonic kidney cell, HEK293-derived human Pepsinogen A5/PGA5 protein Ile16-Ala388, with C-terminal 6-His tag Accession # P0DJD9
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias EC 3.4.23, EC 3.4.23.1, pepsin A, pepsinogen 5, group I (pepsinogen A), pepsinogen A5, PGA3, PGA4, PGA5
Common Name PGA5
Gene Symbol PGA5
Biological Activity Measured by its ability to cleave the fluorogenic peptide substrate, Mca-RPKPVE-Nval-WRK(Dnp)-NH2 . The specific activity is >15,500 pmol/min/μg, as measured under the described conditions.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Ile16-Ala388, with C-terminal 6-His tag Accession # P0DJD9
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane)
Protein PGA5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.