Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Peroxiredoxin 3 Protein
SDP

Catalog No. NBC127993 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC127993 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC127993 Supplier Novus Biologicals™ Supplier No. NBC127993
Only null left
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 63-256 of Human Peroxiredoxin 3 The Recombinant Human Peroxiredoxin 3 Protein is derived from E. coli. The Recombinant Human Peroxiredoxin 3 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Accession Number P30048
For Use With (Application) Bioactivity, Microarray, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 10% glycerol
Gene ID (Entrez) 10935
Molecular Weight (g/mol) 21.5kDa
Name Peroxiredoxin 3 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen Peroxiredoxin 3, 63-256aa, Human, Recombinant, E.coli (NP_006784).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.