Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human PGLYRP1/PGRP-S Protein
SDP

Catalog No. 2590PGB050 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-90P-GB050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 25-90P-GB050 Supplier R&D Systems™ Supplier No. 2590PGB050

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human PGLYRP1/PGRP-S Protein is derived from NS0. The Recombinant Human PGLYRP1/PGRP-S Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number O75594
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in MOPS and NaCl. Reconstitute at 200μg/mL in PBS.
Gene ID (Entrez) 8993
Molecular Weight (g/mol) Observed M.W.: 23-27kDa, reducing conditions, Theoretical M.W.: 20kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human PGLYRP1/PGRP-S protein Gln22-Pro196,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias peptidoglycan recognition protein 1, Peptidoglycan recognition protein short, PGLYRP, PGRPpeptidoglycan recognition protein, PGRPS, PGRP-S, PGRP-SMGC126894, PGRPSMGC126896, TAG7, TNF superfamily, member 3 (LTB)-like (peptidoglycan recognition protein), TNFSF3L
Common Name PGLYRP1/PGRP-S
Gene Symbol PGLYRP1
Biological Activity Measured by its binding ability in a functional ELISA. When peptidoglycan is coated at 1μg/mL (100μL/well), the concentration of Recombinant Human PGLYRP1/PGRP-S that produces 50% optimal binding response is typically 0.75-4.5ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Gln22-Pro196, with a C-terminal 6-His tag
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.