Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human PGLYRP3/PGRP-I alpha Protein

Catalog No. 8507PG050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8507PG050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8507PG050 Supplier R&D Systems™ Supplier No. 8507PG050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human PGLYRP3/PGRP-I alpha Protein is derived from CHO. The Recombinant Human PGLYRP3/PGRP-I alpha Protein has been validated for the following applications: Binding Activity.

Specifications

Accession Number Q96LB9
For Use With (Application) Binding Activity
Gene ID (Entrez) 114771
Molecular Weight (g/mol) Observed M.W.: 34-40kDa, reducing conditions, Theoretical M.W.: 37kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived human PGLYRP3/PGRP-I alpha protein Trp18-His341,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias peptidoglycan recognition protein 3, peptidoglycan recognition protein I alpha, Peptidoglycan recognition protein I-alpha, Peptidoglycan recognition protein intermediate alpha, peptidoglycan recognition protein-I-alpha, PGLYRPIalphaMGC149197, PGRP1A, PGRPI alpha, PGRP-I alpha, PGRPIA, PGRP-Ialpha, PGRPIAPGRP-I-alpha
Common Name PGLYRP3/PGRPIA
Gene Symbol PGLYRP3
Biological Activity Measured by its binding ability in a functional ELISA. When peptidoglycan is coated at 1μg/mL (100μL/well), the concentration of Recombinant Human PGLYRP3/PGRP-I alpha that produces 50% optimal binding response is typically 10-60ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Trp18-His341, with a C-terminal 6-His tag
Purity or Quality Grade >90%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.