Learn More
R&D Systems™ Recombinant Human Phosphodiesterase 4A/PDE4A Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 5141 |
| Molecular Weight (g/mol) | 46 kDa |
| Name | Phosphodiesterase 4A/PDE4A |
| Quantity | 10 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Phosphodiesterase 4A/PDE4A protein Pro331-Met723,with an N-terminal Met and a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Endotoxin Concentration | <0.01 EU per 1μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.