Learn More
Novus Biologicals™ Recombinant Human Phosphoserine phosphatase Protein
Shop All R&D Systems Products

Description
Specifications
Specifications
| Accession Number | P78330 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM HEPES buffer (pH 7.5), 1mM DTT, 100mM KCl2 |
| Gene ID (Entrez) | 5723 |
| Molecular Weight (g/mol) | 25kDa |
| Name | Phosphoserine phosphatase Protein |
| Purification Method | Protein |
| Quantity | 0.1 mg |
| Source | E. Coli |
| Immunogen | Phosphoserine phosphatase Human, Recombinant, E.coli (NP_004568). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.