Learn More
R&D Systems™ Recombinant Human Pin1, HRP-conjugated Protein
Description
Source: E. coli-derived, aa 2-163
Formulation: No Formulation Info in Bulk Inventory
Entrez Gene ID: 5300
Regulatory Status: RUO
Specifications
Specifications
| Formulation | Supplied as a 0.2μM filtered solution in phosphate-buffered saline (PBS) with stabilizers and preservatives. |
| Gene ID (Entrez) | 5300 |
| Name | Pin1 aa 2-163, WW Domain |
| Quantity | 250 μL |
| Storage Requirements | Store the unopened product at 2 - 8° C. Do not use past expiration date. |
| Conjugate | HRP (Horseradish Peroxidase) |
| Recombinant | Recombinant |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.