Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human PKLR Protein

Catalog No. 8569PK050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-69P-K050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 85-69P-K050 Supplier R&D Systems™ Supplier No. 8569PK050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Pyruvate Kinase Isozymes L/R

Measured by its ability to transfer phosphate from phospho(enol)pyruvic acid monosodium salt hydrate (PEP) to adenosine 5'-diphosphate sodium salt (ADP). The specific activity is >25,000 pmol/min/ug, as measured under the described conditions.

Specifications

Accession Number P30613
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and DTT.
Gene ID (Entrez) 5313
Molecular Weight (g/mol) M.W. (Observed): 51-60 kDa, reducing conditions; M.W. (theoretical): 58 kDa
Quantity 50 μg
Source E. coli-derived human PKLR protein Leu47-Ser574, with N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias EC 2.7.1.40, PK1, PK1PKRL, PKL, PKR, Pyruvate kinase 1, pyruvate kinase isozyme R/L, pyruvate kinase isozymes R/L, pyruvate kinase type L, pyruvate kinase, liver and blood cell, pyruvate kinase, liver and RBC, Red cell/liver pyruvate kinase, RPK, R-type/L-type pyruvate kinase
Common Name PKLR
Gene Symbol PKLR
Biological Activity Measured by its ability to transfer phosphate from phospho(enol)pyruvic acid monosodium salt hydrate (PEP) to adenosine 5'-diphosphate sodium salt (ADP). The specific activity is >25,000 pmol/min/ug, as measured under the described conditions.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System E. coli-derived Leu47-Ser574, with N-terminal Met and 6-His tag
Purity or Quality Grade >85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane)
Protein PKLR
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.