Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human PLA2G4A Protein

Catalog No. 6659PL010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6659PL010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 6659PL010 Supplier R&D Systems™ Supplier No. 6659PL010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to hydrolyze 1-Hexadecanoyl-2-(1-pyrene-decanoyl)-sn-glycero-3-phosphocholine. The specific activity is >15 pmol/min/μg, as measured under the described conditions.

Specifications

Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl, TCEP,Brij-35 and Glycerol.
Gene ID (Entrez) 5321
Molecular Weight (g/mol) 86 kDa
Name PLA2G4A
Quantity 10 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human PLA2G4A protein Met1-Ala749,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 80%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.