Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Plasma Kallikrein/KLKB1 Protein

Catalog No. 2497SE010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2497SE010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 2497SE010 Supplier R&D Systems™ Supplier No. 2497SE010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to cleave a flourogenic peptide substrate Pro-Phe-Arg-7-amido-4-methylcoumarin (PFR-AMC). The specific activity is >1,000 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number P03952
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Sodium Acetate and NaCl.
Gene ID (Entrez) 3818
Molecular Weight (g/mol) M.W. (Observed): 80 kDa, reducing conditions; M.W. (theoretical): 70 kDa
Name Plasma Kallikrein/KLKB1
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived human Plasma Kallikrein/KLKB1 protein Gly20-Ala638, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Structural Form Pro and processed forms
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol KLKB1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein Plasma Kallikrein/KLKB1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.