Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human PlGF-4 Protein

Catalog No. 8987PL025CF
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-87P-L025CF 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-87P-L025CF Supplier R&D Systems™ Supplier No. 8987PL025/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human PlGF-4 Protein is derived from HEK293. The Recombinant Human PlGF-4 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P49763-4
For Use With (Application) Bioactivity
Gene ID (Entrez) 5228
Molecular Weight (g/mol) Observed M.W.: 35-40kDa, reducing conditions, Theoretical M.W.: 25kDa
Quantity 25 μg
Source Human embryonic kidney cell,HEK293-derived human PlGF-4 protein Leu19-Arg242
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Common Name PlGF-4
Gene Symbol PGF
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human VEGF R1/Flt-1 Fc Chimera, aa 27-328 is immobilized at 0.5μg/mL, 100μL/well, the concentration of Recombinant Human PlGF-4 that produces 50% of the optimal binding response is approximately 4-24ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Leu19-Arg242
Purity or Quality Grade >85%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.