Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human PlGF Protein

Catalog No. 264PGB010CF
Change view
Click to view available options
Quantity:
10μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
264PGB010CF 10μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 264PGB010CF Supplier R&D Systems™ Supplier No. 264PGB010/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its binding ability in a functional ELISA. When Recombinant Human VEGF R1/Flt-1 Fc Chimera (Catalog # 3516-FL ) is immobilized at 0.5 μg/mL, 100 μL/well, the concentration of Recombinant Human PlGF that produces 50% of the optimal binding response is approximately 0.75-4.5 ng/mL.

Specifications

Accession Number NP_001193941.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Reconstitute at 200μg/mL in 4 mM HCl.
Gene ID (Entrez) 5228
Molecular Weight (g/mol) Observed M.W.: 12-15kDa, reducing conditions, Theoretical M.W.: 15kDa
Quantity 10μg
Source E. coli-derived human PlGF protein Ala21-Arg149
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias D12S1900, PGF, PGFL, placenta growth factor, placental growth factor, placental growth factor, vascular endothelial growth factor-related protein, PlGF, PlGF-2, PLGFplacental growth factor-like, SHGC-10760
Common Name PLGF
Gene Symbol PGF
Biological Activity When Recombinant Human VEGF R1/Flt-1 Fc Chimera is immobilized at 0.5μg/mL, 100μL/well, the concentration of Recombinant Human PlGF that produces 50% of the optimal binding response is approximately 0.15-0.9ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System E. coli-derived Ala21-Arg149
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.