Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human POGLUT1 Protein

Catalog No. 6437GT020
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6437GT020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 6437GT020 Supplier R&D Systems™ Supplier No. 6437GT020
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to hydrolyze UDP-Glucose. The specific activity is >2 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number Q8NBL1
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 56983
Molecular Weight (g/mol) M.W. (Observed): 40-60 kDa, reducing conditions; M.W. (theoretical): 44 kDa
Name Protein O-Glucosyltransferase 1/POGLUT1
Quantity 20 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Protein O-Glucosyltransferase 1/POGLUT1 protein Met1-Leu388, with a C-terminal 6-His tag Accession # Q8NBL1
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol POGLUT1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane)
Protein Protein O-Glucosyltransferase 1/POGLUT1/KTELC1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.