Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Recombinant Human Poliovirus Receptor Protein, Fc Chimera, 100 ug
Description
- Protein Type: Recombinant human Poliovirus Receptor/PVR protein (Fc Chimera Active)
- Expression System: HEK 293 cells
- Protein Range: 21 to 343 aa
- Purity: >95%
- Endotoxin Level: < 1 EU/μg
- Applications: ELISA, Flow Cyt, FuncS, SDS-PAGE
- Biological Activity: Bioactive
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Blue Ice |
| Reactivity | Human |
| Substrate Properties | Protein-based Substrate |
| Buffer | L-Arginine, sodium chloride, 5% trehalose, 0.75% glycine, 0.61% Tris, pH7.4 |
| Target Molecule | Poliovirus Receptor (PVR) |
| Label or Dye | Unconjugated |
| Form | Lyophilized |
| Content And Storage | Store at -20°C. |
| Cross Reactivity | Human |
| Formulation | Lyophilized from L-Arginine, sodium chloride, 5% trehalose, 0.75% glycine, and 0.61% Tris, pH 7.4. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?