Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human POLR3K His Protein
SDP

Catalog No. NBP251772MG Shop All Novus Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP251772MG 0.02 mg
NBP25177101 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP251772MG Supplier R&D Systems™ Supplier No. NBP2517710.02MG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-108 of Human POLR3K The Recombinant Human POLR3K Protein is derived from E. coli. The Recombinant Human POLR3K Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.25mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 0.15M NaCl, 50% glycerol, 2mM DTT, 2mM EDTA
Gene ID (Entrez) 51728
Molecular Weight (g/mol) TMW: 14.7kDa
Quantity 0.02 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias C11, DNA-directed RNA polymerase III subunit K, DNA-directed RNA polymerase III subunit RPC10, DNA-directed RNA polymerases III 12.5 kDa polypeptide, hRPC11, HsC11p, polymerase (RNA) III (DNA directed) polypeptide K (12.3 kDa), polymerase (RNA) III (DNA directed) polypeptide K, 12.3 kDa, RNA polymerase III 12.5 kDa subunit, RNA polymerase III subunit (hRPC11), RNA polymerase III subunit C10, RNA polymerase III subunit C11, RNA polymerase III subunit CII, RPC10, RPC11C11-RNP3, RPC12.5
Common Name POLR3K
Conjugate Unconjugated
Recombinant Recombinant
Product Line Novus
Purity or Quality Grade >85%, by SDS-PAGE
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.