Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human POR/Cytochrome P450 Reductase Protein, CF

Catalog No. 6340PRB020
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-40P-RB020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 63-40P-RB020 Supplier R&D Systems™ Supplier No. 6340PRB020
Add to Cart
Add to Cart
This item is not returnable. View return policy

His-tag

Measured by the reduction of cytochrome c using NADPH as the cofactor. The specific activity is >1800 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number P16435.2
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl, DTT and Glycerol.
Gene ID (Entrez) 5447
Molecular Weight (g/mol) M.W. (Observed): 63-69 kDa, under reducing conditions., M.W. (Theoretical): 71 kDa
Quantity 20 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human POR/Cytochrome P450 Reductase protein Ser62-Ser677 with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias CPR, CYPOR, CYPORFLJ26468, Cytochrome P450 Reductase, DKFZp686G04235, EC 1.6.2.4, NADPH--cytochrome P450 reductase, NADPH-dependent cytochrome P450 reductase, P450 (cytochrome) oxidoreductase, P450R
Conjugate Unconjugated
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein POR/Cytochrome P450 Reductase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.