Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human PP2C alpha/PPM1A Protein
SDP

Catalog No. 4150PP050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4150PP050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 4150PP050 Supplier R&D Systems™ Supplier No. 4150PP050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to dephosphorylate the peptide substrate, DLDVPIPGRFDRRVS(PO3)VAAE(Catalog # ES012 ). The specific activity is >400 nmol/min/mg, as measured under the described conditions.

Specifications

Accession Number NP_808820
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in HEPES, DTT, NaCl and MnCl2.
Gene ID (Entrez) 5494
Molecular Weight (g/mol) M.W. (Observed): 41 kDa, reducing conditions; M.W. (theoretical): 37 kDa
Name PP2C alpha/PPM1A
Quantity 50 μg
Source E. coli-derived human PP2C alpha/PPM1A protein Gly2-Lys324, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Symbol PPM1A
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein PP2C alpha/PPM1A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.