Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human PPP2R2C GST (N-Term) Protein
SDP

Catalog No. NB807668 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB807668 10 μg
NB807473 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB807668 Supplier Novus Biologicals™ Supplier No. H00005522P0110UG
Add to Cart
Add to Cart

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.

Specifications

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 5522
Molecular Weight (g/mol) MolecularWeight-theroretical: 75.9 kDa
Quantity 10 μg
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Gene Alias B55-GAMMA, IMYPNO1, phosphoprotein phosphatase 2A BR gamma regulatory chain, PP2A subunit B isoform B55-gamma, PP2A subunit B isoform gamma, PP2A subunit B isoform PR55-gamma, PP2A subunit B isoform R2-gamma, PP2A, subunit B, B55-gamma isoform, PP2A, subunit B, B-gamma isoform, PP2A, subunit B, PR55-gamma isoform, PP2A, subunit B, R2-gamma isoform, PR55G, protein phosphatase 2, protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), gammaisoform, protein phosphatase 2 (formerly 2A), regulatory subunit B, gamma isoform, protein phosphatase 2, regulatory subunit B, gamma, protein phosphatase 2A1 B gamma subunit, serine/threonine protein phosphatase 2A, 55 KDA regulatory subunit B, gammaisoform, serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B gammaisoform
Gene Symbol PPP2R2C
Research Category Protein Phosphatase
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein PPP2R2C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.