Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human PRL-2/PTP4A2 Protein

Catalog No. 6694PT050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6694PT050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 6694PT050 Supplier R&D Systems™ Supplier No. 6694PT050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to cleave a substrate, p-Nitrophenyl phosphate (pNPP). The specific activity is >0.3 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number Q12974
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in HEPES, NaCl and Betamercaptoethanol.
Gene ID (Entrez) 8073
Molecular Weight (g/mol) M.W. (Observed): 20-22 kDa, reducing conditions; M.W. (theoretical): 20 kDa
Name PRL-2/PTP4A2
Quantity 50 μg
Source E. coli-derived human PRL-2/PTP4A2 protein Asn2-Gln167, with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol PTP4A2
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane)
Protein PRL-2/PTP4A2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.