Learn More
Description
Specifications
Specifications
| Accession Number | NP_116000 |
| For Use With (Application) | ELISA |
| Formulation | 20mM Tris-HCl buffer (pH8.0), 2mM EDTA, 1mM DTT, 10% glycerol |
| Gene ID (Entrez) | 11156 |
| Molecular Weight (g/mol) | 21.6kDa |
| Name | PRL-3/PTP4A3 Protein |
| Purification Method | Protein |
| Immunogen | MGSSHHHHHH SSGLVPRGSH MARMNRPAPV EVSYKHMRFL ITHNPTNATL STFIEDLKKY GATTVVRVCE VTYDKTPLEK DGITVVDWPF DDGAPPPGKV VEDWLSLVKA KFCEAPGSCV AVHCVAGLGR APVLVALALI ESGMKYEDAI QFIRQKRRGA INSKQLTYLE KYRPKQRLRF KDPHTHKTRC CVM |
| Storage Requirements | -80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
