Learn More
R&D Systems™ Recombinant Human Pro-Caspase-3 Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Tris, NaCl, Sucrose and DTT. |
| Gene ID (Entrez) | 836 |
| Molecular Weight (g/mol) | 33 kDa |
| Name | Pro Caspase-3 |
| Quantity | 5 μg |
| Source | E. coli-derived human Caspase-3 protein Met1-His277,with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Pro form |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.